Invitrogen
Manufacturer Code:PA518464
Catalog # PIPA518464
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QPNVDTRQKQ) corresponding to the internal amino acids of VPS25 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Dermal papilla-derived protein 9; ELL-associated protein of 20 kDa; ESCRT-II complex subunit VPS25; hVps25; Vacuolar protein-sorting-associated protein 25; VPS25 vacuolar protein sorting 25 (yeast) HGNC:28122 DERP9 EAP20 MGC10540 vacuolar protein sorting 25 vacuolar protein sorting 25 homolog (S. cerevisiae) FAP20
Gene Aliases: DERP9; EAP20; FAP20; VPS25
UniProt ID: (Human) B2R581, (Rat) P0C0A1
Entrez Gene ID: (Human) 84313, (Rat) 681059
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.