Abnova
Manufacturer Code:PAB6937
Catalog # 89-132-814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human VPS25. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of VPS25.
For Research Use Only
Protein Aliases: Dermal papilla-derived protein 9; DERP9; EAP20; ELL-associated protein of 20 kDa; ESCRT-II complex subunit VPS25; FAP20; hVps25; MGC10540; Vacuolar protein-sorting-associated protein 25
Gene Aliases: DERP9; EAP20; FAP20; VPS25
UniProt ID: (Human) B2R581
Entrez Gene ID: (Human) 84313
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.