Invitrogen
Manufacturer Code:MA537854
Catalog # PIMA537854
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2156 |
Immunogen | A synthesized peptide derived from human VPS18 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 9930024E13Rik hVPS18 KIAA1475 PEP3 vacuolar protein sorting 18 (yeast) vacuolar protein sorting 18 homolog vacuolar protein sorting 18 homolog (S. cerevisiae) vacuolar protein sorting protein 18 vacuolar protein sorting-associated protein 18 homolog VPS18 VPS18 CORVET/HOPS core subunit VPS18 CORVET/HOPS core subunit; hVPS18; vacuolar protein sorting 18 homolog; vacuolar protein sorting protein 18; Vacuolar protein sorting-associated protein 18 homolog; VPS18 CORVET/HOPS core subunit
Gene Aliases: 9930024E13Rik; KIAA1475; PEP3; VPS18
UniProt ID: (Human) Q9P253, (Mouse) Q8R307
Entrez Gene ID: (Human) 57617, (Mouse) 228545, (Rat) 296083
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.