Invitrogen
Manufacturer Code:PA578546
Catalog # PIPA578546
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human VPS18. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hVPS18; PEP3 vacuolar protein sorting 18 homolog (S. cerevisiae); vacuolar protein sorting 18 homolog; vacuolar protein sorting protein 18; Vacuolar protein sorting-associated protein 18 homolog; VPS18 CORVET/HOPS core subunit
Gene Aliases: KIAA1475; PEP3; VPS18
UniProt ID: (Human) Q9P253
Entrez Gene ID: (Human) 57617
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.