Invitrogen
Manufacturer Code:PA537963
Catalog # PIPA537963
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KESQKADEYLREIQ from the internal region of VPS16 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: hVPS16; mVPS16; vacuolar protein sorting 16 homolog; vacuolar protein sorting 16 homolog S. cerevisiae hVPS16 vacuolar protein sorting protein 16 vacuolar protein sorting-associated protein 16 homolog; vacuolar protein sorting protein 16; Vacuolar protein sorting-associated protein 16 homolog; VPS16 CORVET/HOPS core subunit
Gene Aliases: 1810074M16Rik; hVPS16; VPS16
UniProt ID: (Human) Q9H269
Entrez Gene ID: (Human) 64601, (Mouse) 80743, (Rat) 296159
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.