Novus Biologicals
Manufacturer Code:NBP297757AF532
Catalog # NB047433
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human VPS16 fragment. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hVPS16; hVPS16 vacuolar protein sorting 16 homolog (S. cerevisiae) vacuolar protein sorting protein 16 vacuolar protein sorting-associated protein 16 homolog; vacuolar protein sorting 16 homolog; vacuolar protein sorting protein 16; Vacuolar protein sorting-associated protein 16 homolog; VPS16 CORVET/HOPS core subunit
Gene Aliases: hVPS16; VPS16
UniProt ID: (Human) Q9H269
Entrez Gene ID: (Human) 64601
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.