Proteintech
Manufacturer Code:191401AP150UL
Catalog # 50-173-2354
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VPS11 Fusion Protein Ag6227 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: END1 hVPS11 PEP5 RING finger protein 108 RNF108 VPS11; hVPS11; RING finger protein 108; vacuolar protein sorting 11 homolog; Vacuolar protein sorting-associated protein 11 homolog; VPS11 CORVET/HOPS core subunit
Gene Aliases: 1200011A11Rik; END1; HLD12; hVPS11; PEP5; PP3476; RNF108; VPS11
UniProt ID: (Human) Q9H270, (Mouse) Q91W86
Entrez Gene ID: (Human) 55823, (Mouse) 71732, (Rat) 315600
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.