Invitrogen
Manufacturer Code:PA577760
Catalog # PIPA577760
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide CRFHLEIQEEETK corresponding to amino acid residues 25-37 of human VPAC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Helodermin-preferring VIP receptor; PACAP type III receptor; PACAP-R-3; PACAP-R3; Pituitary adenylate cyclase-activating polypeptide type III receptor; Vasoactive intestinal polypeptide receptor 2; vasopressive intestinal peptide receptor 2; VIP and PACAP receptor 2; VIP and PACAP receptor 2 Vasoactive intestinal polypeptide receptor 2 VIPR2 Pituitary Adenylate Cyclase-Activating Polypeptide Type III Receptor; VIP receptor subtype 2; VIP-R-2; VPAC2; VPAC2 receptor
Gene Aliases: C16DUPq36.3; DUP7q36.3; PACAP-R-3; PACAP-R3; VIP-R-2; Vip2; VIP2R; VIPR2; VPAC2; VPAC2R; VPCAP2R
UniProt ID: (Human) P41587, (Rat) P35000, (Mouse) P41588
Entrez Gene ID: (Human) 7434, (Rat) 29555, (Mouse) 22355
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.