Invitrogen
Manufacturer Code:PA534305
Catalog # PIPA534305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 16 amino acid peptide from C-terminus of human VIPR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Helodermin-preferring VIP receptor; PACAP type III receptor; Pituitary adenylate cyclase-activating polypeptide type III receptor; Vasoactive intestinal polypeptide receptor 2; VIP and PACAP receptor 2; VIP-R-2; VIPR2 C16DUPq36.3 DUP7q36.3 PACAP-R-3 Pvr3 Vip type-2 receptor Vip-r2 Vip2 receptor PACAP type III receptor Pacap type-3 PACAP-R3 PACAPR-3 VIP and PACAP receptor 2 VIP-R-2 VIP2R VPAC2 VPCAP2R PACAP3 Vip2 VPAC2R; VPAC2
Gene Aliases: C16DUPq36.3; DUP7q36.3; PACAP-R-3; PACAP-R3; VIP-R-2; VIP2R; VIPR2; VPAC2; VPAC2R; VPCAP2R
UniProt ID: (Human) P41587
Entrez Gene ID: (Human) 7434
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.