Invitrogen
Manufacturer Code:PA527162
Catalog # PIPA527162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 344 and 435 of Human VPAC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Helodermin-preferring VIP receptor; PACAP type III receptor; PACAP-R-3 PACAP-R3 SCZD16 VIP-R-2 VPAC2 VPAC2R VPCAP2R PACAP type III receptor VIP and PACAP receptor 2 helodermin-preferring VIP receptor pituitary adenylate cyclase-activating polypeptide type III receptor vasoactive intestinal polypeptide receptor 2; Pituitary adenylate cyclase-activating polypeptide type III receptor; Vasoactive intestinal polypeptide receptor 2; VIP and PACAP receptor 2; VIP-R-2; VPAC2
Gene Aliases: C16DUPq36.3; DUP7q36.3; PACAP-R-3; PACAP-R3; VIP-R-2; VIP2R; VIPR2; VPAC2; VPAC2R; VPCAP2R
UniProt ID: (Human) P41587
Entrez Gene ID: (Human) 7434
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.