Proteintech
Manufacturer Code:217451AP150UL
Catalog # 50-173-3575
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VNN1 Fusion Protein Ag16501 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HDLCQ8 Pantetheinase Pantetheine hydrolase Tiff66 vanin 1 VNN1; Pantetheinase; Pantetheine hydrolase; Tiff66; Vanin-1; vannin 1; Vascular non-inflammatory molecule 1
Gene Aliases: HDLCQ8; Tiff66; V-1; VNN1
UniProt ID: (Human) O95497, (Mouse) Q9Z0K8
Entrez Gene ID: (Human) 8876, (Mouse) 22361
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.