Invitrogen
Manufacturer Code:PA136031
Catalog # 01-672-221
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Guinea pig |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from the C-terminal of rat VMAT2 conjugated to BSA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Guinea Pig Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC120477 MGC120478 MGC26538 SVAT SVMT VAT2 VMAT2; monoamine neurotransmitter transporter; Monoamine transporter; solute carrier family 18 (vesicular monoamine transporter), member 2; solute carrier family 18 (vesicular monoamine), member 2; Solute carrier family 18 A2 (vesicular monoamine transporter 2); Solute carrier family 18 member 2; solute carrier family 18, member 2; Synaptic vesicular amine transporter; VAT2; vesicle monoamine transporter type 2; vesicle monoamine/H+ antiporter; Vesicular amine transporter 2; vesicular monoamine transporter 2
Gene Aliases: 1110037L13Rik; 9330105E13; MNAT; SLC18A2; SVAT; SVMT; VAT2; VMAT-2; VMAT2
UniProt ID: (Human) Q05940, (Mouse) Q8BRU6, (Rat) Q01827
Entrez Gene ID: (Pig) 100154950, (Human) 6571, (Mouse) 214084, (Rat) 25549
Molecular Function:
amino acid transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.