Novus Biologicals
Manufacturer Code:NBP182783
Catalog # NBP182783
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:PRRTMLRGKSRLNVEWLGYSPGLLLEHRPLLAGRTPRSHRR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MEAXMGC131652 MGC125514 myopathy with excessive autophagy Myopathy with excessive autophagy protein vacuolar ATPase assembly integral membrane protein VMA21 VMA21 vacuolar H+-ATPase homolog (S. cerevisiae) XMEAMGC125516; Myopathy with excessive autophagy protein; Vacuolar ATPase assembly integral membrane protein VMA21
Gene Aliases: MEAX; VMA21; XMEA
UniProt ID: (Human) Q3ZAQ7
Entrez Gene ID: (Human) 203547
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.