Proteintech
Manufacturer Code:219211AP150UL
Catalog # 50-173-3642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VMA21 Fusion Protein Ag16439 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MEAX VMA21 XMEA; Myopathy with excessive autophagy protein; Vacuolar ATPase assembly integral membrane protein VMA21; VMA21 vacuolar H+-ATPase homolog (S. cerevisiae)
Gene Aliases: 2610030H06Rik; AI840175; MEAX; RGD1566155; VMA21; XMEA
UniProt ID: (Human) Q3ZAQ7, (Mouse) Q78T54
Entrez Gene ID: (Human) 203547, (Rat) 501658, (Mouse) 67048
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.