Invitrogen
Manufacturer Code:MA524790
Catalog # PIMA524790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6A6 |
Immunogen | Synthetic peptide corresponding to the C-terminus of human VLDL Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CARMQ1 CHRMQ1 FLJ35024 very low density lipoprotein receptor very low-density lipoprotein receptor VLDL receptor VLDL-R VLDLRCH; non O-glycosylated VLDL-R; Very low-density lipoprotein receptor; VLDL receptor; VLDL-R
Gene Aliases: AA408956; AI451093; AW047288; CAMRQ1; CARMQ1; CHRMQ1; VLDL-R; VLDLR; VLDLRCH
UniProt ID: (Human) P98155, (Mouse) P98156
Entrez Gene ID: (Bovine) 282123, (Human) 7436, (Mouse) 22359, (Rat) 25696
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.