Novus Biologicals
Manufacturer Code:NBP281010
Catalog # NB014083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6A6 |
Immunogen | Synthetic peptide corresponding to the C-terminus of human VLDL Receptor [Swiss-Prot# P98155] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CARMQ1 CHRMQ1 FLJ35024 very low density lipoprotein receptor very low-density lipoprotein receptor VLDL receptor VLDL-R VLDLRCH; Very low-density lipoprotein receptor; VLDL receptor
Gene Aliases: CAMRQ1; CARMQ1; CHRMQ1; VLDL-R; VLDLR; VLDLRCH
UniProt ID: (Human) P98155
Entrez Gene ID: (Human) 7436
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.