Invitrogen
Manufacturer Code:PA547281
Catalog # PIPA547281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse VLDL R Thr25-Ala798 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CARMQ1 CHRMQ1 VLDLRCH VLDL receptor VLDL-R very low-density lipoprotein receptor; Very low-density lipoprotein receptor; VLDL receptor; VLDL-R
Gene Aliases: AA408956; AI451093; AW047288; Vldlr
UniProt ID: (Mouse) P98156
Entrez Gene ID: (Mouse) 22359
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.