Invitrogen
Manufacturer Code:165919050
Catalog # 16-591-9050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Control (Ctrl) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Armenian hamster |
Class | Monoclonal |
Type | Antibody |
Clone | MH5A |
Conjugate | Functional Grade |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Armenian Hamster Monoclonal Antibody
For Research Use Only
Protein Aliases: 4632428N05Rik B7H5 B7-H5 C10orf54 chromosome 10 open reading frame 54 DD1alpha Death Domain1alpha Dies1 differentiation of ESCs 1 GI24 PD-1H PDCD1 homolog PD-H1 platelet receptor Gi24 PP2135 RIKEN cDNA 4632428N05 gene SISP1 sisp-1 Stress-induced secreted protein-1 UNQ730/PRO1412 V-domain Ig suppressor of T cell activation VISTA V-set domain-containing immunoregulatory receptor V-set immunoregulatory receptor Vsir V-type immunoglobulin domain-containing suppressor of T-cell activation; differentiation of ESCs 1; Platelet receptor Gi24; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; V-type immunoglobulin domain-containing suppressor of T-cell activation
Gene Aliases: Dies1; PD-1H; VISTA; Vsir
UniProt ID: (Mouse) Q9D659
Entrez Gene ID: (Mouse) 74048
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.