Invitrogen
Manufacturer Code:PA5113459
Catalog # PIPA5113459
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human V-type immunoglobulin domain-containing suppressor of T-cell activation protein (37-194AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4632428N05Rik B7H5 B7-H5 BOS_24233 C10orf54 C28H10orf54 C4H10orf54 chromosome 10 open reading frame 54 chromosome 28 open reading frame human C10orf54 chromosome 4 open reading frame human C10orf54 DD1alpha Death Domain1alpha Dies1 differentiation of ESCs 1 GI24 PD-1H PDCD1 homolog PD-H1 Platelet receptor Gi24 PP2135 RIKEN cDNA 4632428N05 gene SISP1 Sisp-1 Stress-induced secreted protein-1 UNQ730/PRO1412 V-domain Ig suppressor of T cell activation VISTA V-set domain-containing immunoregulatory receptor V-set immunoregulatory receptor VSIR V-type immunoglobulin domain-containing suppressor of T-cell activation; Death Domain1alpha; differentiation of ESCs 1; PDCD1 homolog; Platelet receptor Gi24; Sisp-1; Stress-induced secreted protein-1; V-domain Ig suppressor of T cell activation; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; V-type immunoglobulin domain-containing suppressor of T-cell activation
Gene Aliases: B7-H5; B7H5; C10orf54; DD1alpha; Dies1; GI24; PD-1H; PP2135; SISP1; UNQ730/PRO1412; VISTA; VSIR
UniProt ID: (Mouse) Q9D659, (Human) Q9H7M9
Entrez Gene ID: (Mouse) 74048, (Human) 64115
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.