R&D Systems
Manufacturer Code:FAB5416P025
Catalog # FAB5416P025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 476031 |
Immunogen | NS0 mouse myeloma cell line transfected with human VIP R2 Glu24-Ile438 Accession # P41587 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ16511 Helodermin-preferring VIP receptor PACAP type III receptor PACAPR3 PACAP-R3 PACAP-R-3 Pituitary adenylate cyclase-activating polypeptide type III receptor vasoactive intestinal peptide receptor 2 vasoactive intestinal polypeptide receptor 2 VIP R2 VIP2R VIP-R-2 VPAC2 VPCAP2R; Helodermin-preferring VIP receptor; PACAP type III receptor; Pituitary adenylate cyclase-activating polypeptide type III receptor; Vasoactive intestinal polypeptide receptor 2; VIP and PACAP receptor 2; VIP-R-2; VPAC2
Gene Aliases: C16DUPq36.3; DUP7q36.3; PACAP-R-3; PACAP-R3; VIP-R-2; VIP2R; VIPR2; VPAC2; VPAC2R; VPCAP2R
UniProt ID: (Human) P41587
Entrez Gene ID: (Human) 7434
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.