Abnova
Manufacturer Code:PAB27706
Catalog # 89-129-223
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acid at cytoplasmic domain of human VIPR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of VIPR1.
For Research Use Only
Protein Aliases: FLJ41949; HVR1; II; PACAP type II receptor; PACAP-R-2; pituitary adenylate cyclase activating polypeptide receptor, type II; Pituitary adenylate cyclase-activating polypeptide type II receptor; RDC1; type 1 vasoactive intestinal peptide receptor; VAPC1; Vasoactive intestinal polypeptide receptor 1; VIP and PACAP receptor 1; VIP receptor, type I; VIP-R-1; VIPR; VIRG; VPAC1; VPAC1 receptor; VPCAP1R
Gene Aliases: HVR1; II; PACAP-R-2; PACAP-R2; RDC1; V1RG; VAPC1; VIP-R-1; VIPR; VIPR1; VIRG; VPAC1; VPAC1R; VPCAP1R
UniProt ID: (Human) P32241
Entrez Gene ID: (Human) 7433
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.