OriGene
Manufacturer Code:TA806851
Catalog # 50-168-1284
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F3 |
Immunogen | Full length human recombit protein of human VIP produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Intestinal peptide PHM-27; Intestinal peptide PHV-42; Peptide histidine methioninamide 27; Peptide histidine valine 42; PHM27; prepro-VIP; Vasoactive intestinal peptide; Vasoactive intestinal polypeptide; VIP; VIP peptides
Gene Aliases: PHM27; VIP
UniProt ID: (Human) P01282
Entrez Gene ID: (Human) 7432
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.