Invitrogen
Manufacturer Code:ABS0230202
Catalog # ABA-230202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2 |
Immunogen | Vasoactive intestinal peptide. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Intestinal peptide PHI-27; Intestinal peptide PHM-27; Intestinal peptide PHV-42; Peptide histidine isoleucinamide 27; Peptide histidine methioninamide 27; Peptide histidine valine 42; prepro-VIP; Vasoactive intestinal peptide; Vasoactive intestinal polypeptide; VIP; VIP peptides; VIP-R-1 Pituitary adenylate cyclase-activating polypeptide type II receptor PACAP type II receptor PACAP-R-2
Gene Aliases: PHI; PHM27; VIP; vip/phi27
UniProt ID: (Bovine) P81401, (Human) P01282, (Rat) P01283
Entrez Gene ID: (Bovine) 280956, (Human) 7432, (Pig) 100500718, (Rat) 117064
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.