OriGene
Manufacturer Code:CF801117
Catalog # 50-166-5457
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 210-466 of human VIM produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: epididymis luminal protein 113; Vimentin
Gene Aliases: CTRCT30; HEL113; VIM
UniProt ID: (Human) P08670
Entrez Gene ID: (Human) 7431
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.