OriGene
Manufacturer Code:CF803236
Catalog # 50-166-6389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-188 of human VIL1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: D2S1471 VIL; Villin-1
Gene Aliases: D2S1471; VIL; VIL1
UniProt ID: (Human) P09327
Entrez Gene ID: (Human) 7429
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.