R&D Systems
Manufacturer Code:AF6847SP
Catalog # AF6847SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human VIAAT/SLC32A1 Ala2-Phe133 Accession # Q9H598 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: bA122O1.1 hVIAAT SLC32A1 solute carrier family 32 (GABA vesicular transporter) member 1 Solute carrier family 32 member 1 Vesicular GABA transporter vesicular inhibitory amino acid transporter VGAT VGATGABA and glycine transporter VIAAT; GABA and glycine transporter; hVIAAT; solute carrier family 32 (GABA vesicular transporter), member 1; Solute carrier family 32 member 1; Vesicular GABA transporter; Vesicular inhibitory amino acid transporter
Gene Aliases: SLC32A1; VGAT; VIAAT
UniProt ID: (Human) Q9H598
Entrez Gene ID: (Human) 140679
Molecular Function: amino acid transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.