Novus Biologicals
Manufacturer Code:NBP133099
Catalog # NBP133099
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full length human DUSP3 Recombinant protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dual specificity phosphatase 3 dual specificity protein phosphatase 3 Dual specificity protein phosphatase VHR EC 3.1.3.16 EC 3.1.3.48 Vaccinia H1-related phosphatase vaccinia virus phosphatase VH1-related VHRserine/threonine specific protein phosphatase; Dual specificity protein phosphatase 3; Dual specificity protein phosphatase VHR; serine/threonine specific protein phosphatase; Vaccinia H1-related phosphatase; vaccinia virus phosphatase VH1-related; VHR
Gene Aliases: DUSP3; VHR
UniProt ID: (Human) P51452
Entrez Gene ID: (Human) 1845
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.