Novus Biologicals
Manufacturer Code:NBP259331AF647
Catalog # NB259331647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S34-34 |
Immunogen | Fusion protein amino acids 546-588 (cytoplasmic C-terminus) of rat VGlut3 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter) member 8; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8; solute carrier family 17 (vesicular glutamate transporter), member 8; Solute carrier family 17 member 8; Vesicular glutamate transporter 3; VGluT3
Gene Aliases: DFNA25; SLC17A8; VGLUT3
UniProt ID: (Human) Q8NDX2
Entrez Gene ID: (Human) 246213
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.