Novus Biologicals
Manufacturer Code:NBP1259730.025ML
Catalog # NB018469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the c-terminal region of mouse VGLUT3 conjugated to an immunogenic carrier protein was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter) member 8; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8; solute carrier family 17 (vesicular glutamate transporter), member 8; Solute carrier family 17 member 8; Vesicular glutamate transporter 3; VGluT3
Gene Aliases: DFNA25; SLC17A8; VGLUT3
UniProt ID: (Human) Q8NDX2
Entrez Gene ID: (Human) 246213
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.