Invitrogen
Manufacturer Code:PA585784
Catalog # PIPA585784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of mouse vGluT3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC042593 antibody Slc17a8 antibody solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter) member 8 antibody; Solute carrier family 17 member 8; Vesicular glutamate transporter 3; vesicular glutamate transporter-3; VGluT3
Gene Aliases: BC042593; Slc17a8; Vglut3
UniProt ID: (Mouse) Q8BFU8
Entrez Gene ID: (Mouse) 216227
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.