Novus Biologicals
Manufacturer Code:NBP259329B
Catalog # NBP259329B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S28-9 |
Immunogen | Fusion protein amino acids 493-560 (cytoplasmic C-terminus) of rat VGlut1 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: BNPIbrain-specific Na-dependent inorganic phosphate cotransporter Brain-specific Na(+)-dependent inorganic phosphate cotransporter member 7 solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter) Solute carrier family 17 member 7 VGluT1 VGLUT1vesicular glutamate transporter 1; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; solute carrier family 17 (vesicular glutamate transporter), member 7; Solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGluT1
Gene Aliases: BNPI; SLC17A7; VGLUT1
UniProt ID: (Human) Q9P2U7
Entrez Gene ID: (Human) 57030
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.