R&D Systems
Manufacturer Code:MAB9054
Catalog # MAB9054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 732607 |
Immunogen | <em>E.coli</em>-derived recombinant human VGLUT/SLC17A7 Asn93-Thr115 Accession # Q9P2U7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BNPI BNPIbrain-specific Na-dependent inorganic phosphate cotransporter Brain-specific Na(+)-dependent inorganic phosphate cotransporter member 7 SLC17A7 solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter) Solute carrier family 17 member 7 VGluT1 VGLUT1vesicular glutamate transporter 1; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; solute carrier family 17 (vesicular glutamate transporter), member 7; Solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGluT1
Gene Aliases: BNPI; SLC17A7; VGLUT1
UniProt ID: (Human) Q9P2U7
Entrez Gene ID: (Human) 57030
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.