Invitrogen
Manufacturer Code:PA588830
Catalog # PIPA588830
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 461-560 of human SLC17A7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain-specific Na(+)-dependent inorganic phosphate cotransporter; Solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGluT1
Gene Aliases: 2900052E22Rik; AI851913; Bnpi; Slc17a7; Vglut1
UniProt ID: (Mouse) Q3TXX4
Entrez Gene ID: (Mouse) 72961
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.