Novus Biologicals
Manufacturer Code:NB10018370.05MG
Catalog # NB100183705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Radioimmune Assays (RIA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 residue synthetic peptide derived from C-terminal domain of rat VGLUT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BNPIbrain-specific Na-dependent inorganic phosphate cotransporter Brain-specific Na(+)-dependent inorganic phosphate cotransporter member 7 solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter) Solute carrier family 17 member 7 VGluT1 VGLUT1vesicular glutamate transporter 1; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; solute carrier family 17 (vesicular glutamate transporter), member 7; Solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGluT1
Gene Aliases: BNPI; SLC17A7; VGLUT1
UniProt ID: (Human) Q9P2U7
Entrez Gene ID: (Human) 57030
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.