Invitrogen
Manufacturer Code:PA595097
Catalog # PIPA595097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human VGF different from the related rat and mouse sequences by one amino acid. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antimicrobial peptide VGF[554-577]; NERP-1; NERP-2; neuro-endocrine specific protein VGF; Neuroendocrine regulatory peptide-1; Neuroendocrine regulatory peptide-2; Neurosecretory protein VGF; Neurosecretory protein VGF Neuroendocrine regulatory peptide-1 NERP-1 Neuroendocrine regulatory peptide-2 NERP-2 Antimicrobial peptide VGF&lsqb554-577] VGF; VGF(180-194); VGF(24-63); VGF(375-407); VGF-derived peptide AQEE-30; VGF-derived peptide HFHH-10; VGF-derived peptide LQEQ-19; VGF-derived peptide TLQP-11; VGF-derived peptide TLQP-21; VGF-derived peptide TLQP-30; VGF-derived peptide TLQP-62; VGF8a protein
Gene Aliases: Gm1052; SCG7; SgVII; VGF
UniProt ID: (Human) Q9UDW8, (Rat) P20156, (Mouse) Q0VGU4
Entrez Gene ID: (Human) 7425, (Rat) 29461, (Mouse) 381677
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.