Invitrogen
Manufacturer Code:PA584900
Catalog # PIPA584900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human VGF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antimicrobial peptide VGF[554-577]; NERP-1; NERP-2; neuro-endocrine specific protein VGF; Neuroendocrine regulatory peptide-1; Neuroendocrine regulatory peptide-2; Neurosecretory protein VGF; SCG7 SgVII; VGF-derived peptide TLQP-21; VGF-derived peptide TLQP-62
Gene Aliases: SCG7; SgVII; VGF
UniProt ID: (Human) Q9UDW8
Entrez Gene ID: (Human) 7425
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.