R&D Systems
Manufacturer Code:FAB3492A
Catalog # FAB3492A
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 54733 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human VEGF R3/Flt-4 Tyr25-Ile776 Accession # P35916 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.1 Flt-4 FLT4 fms-related tyrosine kinase 4 LMPH1A PCLFLT41 soluble VEGFR3 variant 1 soluble VEGFR3 variant 2 soluble VEGFR3 variant 3 Tyrosine-protein kinase receptor FLT4 vascular endothelial growth factor receptor 3 VEGF R3 VEGFR-3 VEGFR3Fms-like tyrosine kinase 4; FLT-4; Fms-like tyrosine kinase 4; fms-related tyrosine kinase 4; Tyrosine-protein kinase receptor FLT4; Vascular endothelial growth factor receptor 3; VEGFR-3
Gene Aliases: FLT-4; FLT4; FLT41; LMPH1A; PCL; VEGFR-3; VEGFR3
UniProt ID: (Human) P35916
Entrez Gene ID: (Human) 2324
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.