Novus Biologicals
Manufacturer Code:NB100530
Catalog # NB100530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to a C-terminal region of the mouse VEGF Receptor 2 protein (between residues 1300-1367). [Swiss-Prot# P35918] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD309; CD309 CD309 antigen EC 2.7.10 EC 2.7.10.1 Fetal liver kinase 1 fetal liver kinase-1 FLK-1 FLK1tyrosine kinase growth factor receptor Kinase insert domain receptor kinase insert domain receptor (a type III receptor tyrosine kinase) Protein-tyrosine kinase receptor flk-1 soluble VEGFR2 vascular endothelial growth factor receptor 2 VEGFR VEGFR2 VEGFR-2; Fetal liver kinase 1; fetal liver kinase-1; FLK-1; KDR; Kinase insert domain receptor; kinase insert domain receptor (a type III receptor tyrosine kinase); Protein-tyrosine kinase receptor flk-1; soluble VEGFR2; tyrosine kinase growth factor receptor; Vascular endothelial growth factor receptor 2; VEGFR-2
Gene Aliases: CD309; FLK1; KDR; VEGFR; VEGFR2
UniProt ID: (Human) P35968
Entrez Gene ID: (Human) 3791
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.