Novus Biologicals
Manufacturer Code:NBP118631
Catalog # NBP118631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide: GEHCNKKAVFSRISKFKSTR NDSTTQSNVKH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.1 FLT FLT-1 Fms-like tyrosine kinase 1 fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor) FRT Tyrosine-protein kinase FRT Tyrosine-protein kinase receptor FLT vascular endothelial growth factor receptor 1 Vascular permeability factor receptor VEGFR1 VEGFR-1; FLT; FLT-1; Fms-like tyrosine kinase 1; fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); Tyrosine-protein kinase FRT; Tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; Vascular permeability factor receptor; VEGFR-1
Gene Aliases: FLT; FLT-1; FLT1; FRT; VEGFR-1; VEGFR1
UniProt ID: (Human) P17948
Entrez Gene ID: (Human) 2321
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.