Proteintech
Manufacturer Code:136871AP150UL
Catalog # 50-172-8936
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VEGFR-1/FLT-1 Fusion Protein Ag4558 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Embryonic receptor kinase 2; FLT; FLT FLT 1 FLT1 FLT-1 Fms like tyrosine kinase 1 FRT sflt1 Tyrosine protein kinase FRT VEGF receptor 1 VEGFR 1 VEGFR1 VEGFR-1; FLT-1; FMS-like tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); Tyrosine-protein kinase FRT; Tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; vascular endothelial growth factor receptor-1; Vascular permeability factor receptor; VEGFR-1
Gene Aliases: AI323757; Emrk2; FLT; FLT-1; FLT1; FRT; sFlt1; VEGFR-1; VEGFR1
UniProt ID: (Human) P17948, (Mouse) P35969
Entrez Gene ID: (Human) 2321, (Mouse) 14254, (Rat) 54251
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.