Invitrogen
Manufacturer Code:PA580220
Catalog # PIPA580220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat VEGFB recombit protein (Position: P22-A207). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Vascular endothelial growth factor B; Vascular endothelial growth factor B VEGF-B VEGF-related factor VRF Vegfb Vrf; VEGF-B; VEGF-related factor; VRF
Gene Aliases: VEGF-B; Vegfb; Vrf
UniProt ID: (Rat) O35485, (Mouse) P49766
Entrez Gene ID: (Rat) 89811, (Mouse) 22340
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.