Invitrogen
Manufacturer Code:PA544952
Catalog # PIPA544952
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human VEGFB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: flt-1/VEGFR1 ligand; Vascular endothelial growth factor B; Vascular endothelial growth factor B VEGFL VRF; VEGF-B; VEGF-related factor; VRF
Gene Aliases: VEGF-B; VEGFB; VEGFL; VRF
UniProt ID: (Human) P49765, (Bovine) Q9XS49, (Rat) O35485, (Mouse) P49766
Entrez Gene ID: (Human) 7423, (Bovine) 282121, (Horse) 100055238, (Dog) 476036, (Pig) 100521837, (Rat) 89811, (Mouse) 22340
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.