Abnova
Manufacturer Code:PAB13528
Catalog # 89-116-724
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to mouse Vegfb. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere.<br />Aliquot to avoid repeated freezing and thawing. |
Rat polyclonal antibody raised against recombinant Vegfb.
For Research Use Only
Protein Aliases: Vascular endothelial growth factor B; VEGF-B; VEGF-related factor; VRF
Gene Aliases: VEGF-B; Vegfb; Vrf
UniProt ID: (Mouse) P49766
Entrez Gene ID: (Mouse) 22340
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.