Abnova
Manufacturer Code:MAB5374
Catalog # 89-112-234
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SP28 |
Immunogen | A synthetic peptide corresponding to N-terminus of human VEGFA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit monoclonal antibody raised against synthetic peptide of VEGFA.
For Research Use Only
Protein Aliases: MGC70609; Vascular endothelial growth factor A; vascular endothelial growth factor A121; vascular endothelial growth factor A165; Vascular permeability factor; VEGF; VEGF-A; VPF
Gene Aliases: MVCD1; VEGF; VEGFA; VPF
UniProt ID: (Human) P15692
Entrez Gene ID: (Human) 7422
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.