Abnova
Manufacturer Code:PAB13526
Catalog # 89-116-723
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to mouse Vegfa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. After reconstitution with deionized water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant Vegfa.
For Research Use Only
Protein Aliases: vascular endothelial growth factor 165b; Vascular endothelial growth factor A; Vascular permeability factor; Vegf; VEGF-A; Vegf120; Vegf164; Vegf188; VPF
Gene Aliases: Vegf; Vegfa; Vpf
UniProt ID: (Mouse) Q6GT23
Entrez Gene ID: (Mouse) 22339
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.