Invitrogen
Manufacturer Code:MA535280
Catalog # PIMA535280
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0363 |
Immunogen | A synthesized peptide derived from human VEGF Receptor 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AI323757 Embryonic receptor kinase 2 Emrk2 FLT FLT1 FLT-1 Fms-like tyrosine kinase 1 FRT i21 sFlt1 soluble VEGFR 1 sVEGFR 1 Tyrosine-protein kinase FRT Tyrosine-protein kinase receptor FLT Vascular endothelial growth factor receptor 1 Vascular permeability factor receptor VEGF R1 Vegfr1 VEGFR-1; Embryonic receptor kinase 2; FLT-1; FMS-like tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); Tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; vascular endothelial growth factor receptor-1; vascular permeability factor receptor; VEGFR-1
Gene Aliases: AI323757; Emrk2; Flt; Flt-1; Flt1; sFlt1; VEGFR-1; VEGFR1
UniProt ID: (Mouse) P35969
Entrez Gene ID: (Mouse) 14254, (Rat) 54251
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.