Invitrogen
Manufacturer Code:PA116612
Catalog # APPA6612
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal region of the murine VEGFR1 (residues 800-900): 87&percnt identity to the VEGFR-2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD309; Embryonic receptor kinase 2; Fetal liver kinase 1; fetal liver kinase-1; FLK-1; FLT; FLT-1; Fms-like tyrosine kinase 1; fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); KDR; Kinase insert domain receptor; kinase insert domain receptor (a type III receptor tyrosine kinase); Protein-tyrosine kinase receptor flk-1; soluble VEGFR2; tyrosine kinase growth factor receptor; Tyrosine-protein kinase FRT; Tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; Vascular endothelial growth factor receptor 2; vascular endothelial growth factor receptor-1; Vascular permeability factor receptor; VEGFR-1; VEGFR-2; VEGFR1 VEGFR 1 Vascular permeability factor receptor Tyrosine-protein kinase receptor FLT Flt-1 Tyrosine-protein kinase FRT VEGFR2
Gene Aliases: AI323757; CD309; Emrk2; FLK1; FLT; FLT-1; FLT1; FRT; KDR; sFlt1; VEGFR; VEGFR-1; VEGFR1; VEGFR2
UniProt ID: (Human) P17948, (Mouse) P35969, (Human) P35968
Entrez Gene ID: (Human) 2321, (Mouse) 14254, (Human) 3791
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.