Invitrogen
Manufacturer Code:MA116626
Catalog # AMPA6626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | VG76e |
Immunogen | Human VEGF189 expressed in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: angiogenic factor; unnamed protein product; vascular endothelial growth factor; vascular endothelial growth factor (VEGF) precursor; vascular endothelial growth factor 164; Vascular endothelial growth factor A; vascular endothelial growth factor A121; vascular endothelial growth factor A165; Vascular Endothelial Growth Factor Vascular Permeability Factor VEGFA VPF; Vascular permeability factor; VEGF; VEGF-A; VEGF182; VEGF188; VPF
Gene Aliases: eVEGF120; eVEGF164; MVCD1; VEGF; VEGF-A; VEGFA; VPF
UniProt ID: (Bovine) P15691, (Human) P15692, (Sheep) P50412, (Pig) P49151
Entrez Gene ID: (Bovine) 281572, (Human) 7422, (Sheep) 443103, (Pig) 397157
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.