Invitrogen
Manufacturer Code:MA523769
Catalog # PIMA523769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 123704 |
Immunogen | Mouse myeloma cell line NS0-derived recombit rat VEGF164 Ala27-Arg190 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Vascular endothelial growth factor A; Vascular Endothelial Growth Factor Vascular Permeability Factor VEGFA VPF; Vascular permeability factor; VEGF-A; VPF
Gene Aliases: Vegf; VEGF-A; VEGF164; Vegfa; VPF
UniProt ID: (Rat) P16612
Entrez Gene ID: (Rat) 83785
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.