R&D Systems
Manufacturer Code:AF493SP
Catalog # AF493SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <em>S. frugiperda</em> insect ovarian cell line <em>Sf</em> 21-derived recombinant mouse VEGF 164 Ala27-Arg190 Accession # AAA40547 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: MVCD1 VAS vascular endothelial growth factor A Vascular permeability factor Vasculotropin VEGFA VEGF-A VEGFMGC70609 VPF VPFvascular endothelial growth factor; Vascular endothelial growth factor A; vascular endothelial growth factor A121; vascular endothelial growth factor A165; Vascular permeability factor; VEGF-A; VPF
Gene Aliases: MVCD1; VEGF; VEGFA; VPF
UniProt ID: (Human) P15692
Entrez Gene ID: (Human) 7422
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.